Cole-Parmer VapLock™ Plug, PFA, Luer Plug x 1/4-28 UNF(M) Flat Bottom; 1/Ea
Representative image only.
Cole-Parmer VapLock™ Plug, PFA, Luer Plug x 1/4-28 UNF(M) Flat Bottom; 1/Ea
Cole-Parmer | Item: 1203120
Use plugs to seal unused ports for a leak-resistant system
₹790.00INR / Each
₹790.00INR / Each
Product Description
Key Features
  • Seal unused manifold and bottle cap ports
More About this Item
Bottom-sealing UNF thread options include 1/4-28 and 5/16-24, and do not require PTFE tape for leak-resistant connections.
Specifications
  • Fitting MaterialPFA
  • Connection TypeUNF(M)
  • Thread size UNF(M) (port 1) (InchShort)1/4"-28
  • ColorWhite
  • DescriptionPlug, PFA, Luer Plug x 1/4-28 UNF(M) Flat Bottom; 1/Ea