Cole-Parmer Standard Benchtop Chilling/Heating Block, 100-230 VAC
Representative image only.
Cole-Parmer Standard Benchtop Chilling/Heating Block, 100-230 VAC
Cole-Parmer | Item: 4417500
Chill, freeze, heat, or mix biological samples at your work station
₹434,000.00INR / Each
₹434,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Standard Benchtop Chilling/Heating Block, 100-230 VAC
Cole-Parmer Standard Benchtop Chilling/Heating Block, 100-230 VAC
4417500
₹434,000.00INR / Each
+ + + + =
Total Price 1127500.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Chiller/heater module, power cord, instruction manual, and a 12 VDC power supply. Order cover and aluminum blocks separately.
Key Features
  • Store 5 programs in memory for precisely timed and repeatable routines
  • Choose blocks for centrifuge tubes, assay plates, PCR tubes & plates, and more
  • Two-block unit offers two different or same temperatures
  • 1 year warranty
More About this Item
Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM® compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100°C with ±1°C digital control.
Standard Chilling/Heating Block is ideal for a variety of applications such as storing enzymes or DNA libraries, maintaining 14°C for ligation reactions, and incubating or deactivating enzyme reactions.
1 Year WarrantyCanadian Standards AssociationUL Listed.
Specifications
  • Max Temperature (° F)212
  • Max Temperature (° C)100
  • Power (VAC)100 to 230
  • DescriptionStandard Benchtop Chilling/Heating Block, 100-230 VAC