Cole-Parmer Benchtop Chilling/Heating/Mixing Blocks
Chill, freeze, heat, or mix biological samples at your work station
  • Store 5 programs in memory for precisely timed and repeatable routines
  • Choose blocks for centrifuge tubes, assay plates, PCR tubes & plates, and more
  • Two-block unit offers two different or same temperatures
Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM® compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100°C with ±1°C digital control.
287,000.00 - ₹666,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
100 to 230 Standard Benchtop Chilling/Heating Block, 100-230 VAC
₹287,000.00INR / Each
COMPARE
100 to 230 Orbital Chilling/Heating Block, 100 to 230 VAC
₹592,000.00INR / Each
COMPARE
100 to 230 Programmable Orbital Mixing/Chilling/Heating Block, 100 to 230V
₹666,000.00INR / Each
COMPARE
100 to 230 Two-Position Chilling/Heating Block, 100 VAC
₹494,000.00INR / Each