Cole-Parmer Orbital Chilling/Heating Block, 100 to 230 VAC

Representative image only.

Cole-Parmer Orbital Chilling/Heating Block, 100 to 230 VAC

Cole-Parmer | Item: 4417502

Chill, freeze, heat, or mix biological samples at your work station

₹927,000.00INR / Each

₹927,000.00INR / Each

Buy It Together

Current Item

Cole-Parmer Orbital Chilling/Heating Block, 100 to 230 VAC
Cole-Parmer Orbital Chilling/Heating Block, 100 to 230 VAC
4417502

₹927,000.00INR / Each
+ + + + =
Total Price 1620500.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Chiller/heater module, power cord, instruction manual, and a 12 VDC power supply. Order cover and aluminum blocks separately.

Key Features

  • Store 5 programs in memory for precisely timed and repeatable routines
  • Choose blocks for centrifuge tubes, assay plates, PCR tubes & plates, and more
  • Two-block unit offers two different or same temperatures
  • 1 year warranty

More About this Item

Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM® compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100°C with ±1°C digital control.

Orbital Mixing Chilling/Heating Block combines shaking, heating, and cooling all in one instrument with a built-in orbital mixer with speeds settable from 200 to 1000 rpm.

Conformite Europeenne1 Year WarrantyCanadian Standards Association
Specifications
  • Power (VAC)100 to 230
  • DescriptionOrbital Chilling/Heating Block, 100 to 230 VAC
  • Warranty1 year
Reviews
Q&A