Cole-Parmer Aluminum blocks, Flat Bottom for 96/384 Well Assay Plates
Representative image only.
Cole-Parmer Aluminum blocks, Flat Bottom for 96/384 Well Assay Plates
Cole-Parmer | Item: 4417536
₹146,000.00INR / Each
₹146,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Aluminum blocks, Flat Bottom for 96/384 Well Assay Plates
Cole-Parmer Aluminum blocks, Flat Bottom for 96/384 Well Assay Plates
4417536
₹146,000.00INR / Each
+ +
Cole-Parmer Cover, Plastic; for Aluminum Blocks
4417550
₹22,700.00INR / Each
+ =
Total Price 625600.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Change the capacity of your dry block heater with interchangeable blocks.
1 Year Warranty
Specifications
  • Block Number Of Openings1
  • Combination Block -Tube / Vial Size Accepted96/384-well PCR plate
  • Width (in)2 7/8
  • Length (in)4 3/8
  • DescriptionAluminum blocks, Flat Bottom for 96/384 Well Assay Plates