Cole-Parmer CB-200D Block Chiller/Heater; 100-230 VAC, 50/60 Hz

Representative image only.

Cole-Parmer CB-200D Block Chiller/Heater; 100-230 VAC, 50/60 Hz

Cole-Parmer | Item: 3662017

Safely maintain biological samples

₹222,000.00INR / Each

for an additional
₹32,100.00INR / Each
for an additional
₹44,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹222,000.00INR / Each

Buy It Together

Current Item

Cole-Parmer CB-200D Block Chiller/Heater; 100-230 VAC, 50/60 Hz
Cole-Parmer CB-200D Block Chiller/Heater; 100-230 VAC, 50/60 Hz
3662017

₹222,000.00INR / Each
+ + + + =
Total Price 768900.00 ADD 5 ITEMS TO CART
Product Description

Includes

Lid and power cords with plug (US, UK, and EU).

Key Features

  • Designed for maintaining biological samples safely between 4 and 37°C
  • Alarm sounds when set temperature is reached and elapsed time ends
  • Five-digit LED display can be set for either °C or °F
  • Choose from a wide range of interchangeable aluminum block inserts
  • 3 year warranty
  • Previous part number FBLOCKICE

More About this Item

This benchtop block chiller/heater is ideal for incubating sample tubes at temperatures ranging from 32 to 104°F (0 to 40°C). The Isotemp® temperature control ensures all contents are kept at an even temperature, whether cold or warm. Integrated countdown/count-up timer with audible alarm allows up to nine days to be programmed and will not start until a stable temperature has been reached.

Use with Dri-Block® aluminum inserts to maintain temperature stability of ±0.2°C. Unit holds two single-width inserts or one double-width inserts. A dark plastic lid protects light-sensitive reagents as well as helping to maintain the sample temperature.

Please Note: Effective June 2022, the Techne Cooling/Heating Block Model number FBLOCKICE has now been rebranded Cole-Parmer under Model number CB-200D. All the same great features and specifications, just with a new and improved look.

3 Year WarrantyConformite Europeenne
Specifications
  • Number Of Blocks2
  • Uniformity (° F)±0.36°F (0.2°C)
  • Max Temperature (° F)104
  • Max Temperature (° C)40
  • Power (VAC)100 to 230
  • Block TypeChilling/Heating Blocks
  • Height (in)8 7/8
  • Width (in)9 7/16
  • Depth (in)7 43/64
  • DescriptionBlock Chiller/Heater; 100-230 VAC, 50/60 Hz
  • Warranty3 Years
Reviews
Q&A