Cole-Parmer Programmable Orbital Mixing/Chilling/Heating Block, 100 to 230V
Representative image only.
Cole-Parmer Programmable Orbital Mixing/Chilling/Heating Block, 100 to 230V
Cole-Parmer | Item: 4417504
Chill, freeze, heat, or mix biological samples at your work station
₹1,033,000.00INR / Each
₹1,033,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Programmable Orbital Mixing/Chilling/Heating Block, 100 to 230V
Cole-Parmer Programmable Orbital Mixing/Chilling/Heating Block, 100 to 230V
4417504
₹1,033,000.00INR / Each
+ + + + =
Total Price 1726500.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Chiller/heater module, power cord, instruction manual, and a 12 VDC power supply. Order cover and aluminum blocks separately.
Key Features
  • Store 5 programs in memory for precisely timed and repeatable routines
  • Choose blocks for centrifuge tubes, assay plates, PCR tubes & plates, and more
  • Two-block unit offers two different or same temperatures
  • 1 year warranty
More About this Item
Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM® compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100°C with ±1°C digital control.
Programmable Orbital Mixing Chilling/Heating Block is fully programmable with built-in orbital mixer. This model can store 5 programs in memory of as many as 10 steps per program where each step can be a temperature, time and mixing speed. Each program can be repeated from 1 to 99 times automatically.
Conformite Europeenne1 Year WarrantyCanadian Standards Association
Specifications
  • Power (VAC)100 to 230
  • DescriptionProgrammable Orbital Mixing/Chilling/Heating Block, 100 to 230V