Cole-Parmer Replacement tube for deluxe ultraviolet viewing cabinet, 312 nm

Representative image only.

Cole-Parmer Replacement tube for deluxe ultraviolet viewing cabinet, 312 nm

Cole-Parmer | Item: 0981563

₹29,300.00INR / Each

₹29,300.00INR / Each

Buy It Together

Current Item

Cole-Parmer Replacement tube for deluxe ultraviolet viewing cabinet, 312 nm
Cole-Parmer Replacement tube for deluxe ultraviolet viewing cabinet, 312 nm
0981563

₹29,300.00INR / Each
+ + + + + +
Ultraviolet Crosslinker Replacement Tube; 302 nm
9760751

₹17,100.00INR / Each
=
Total Price 399000.00 ADD 7 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Keep your ultraviolet equipment running and dependable with replacement bulbs and accessories.

Product may contain trace amounts of mercury; cautions should be used in breakage, cleanup, shipping and/or disposal.

Specifications
  • DescriptionReplacement tube for deluxe ultraviolet viewing cabinet, 312 nm
Reviews
Q&A