Pen-Ray UV Lamps
Ideal for UV testing through small apertures
  • Economically calibrate your photometric and fluorometric instruments
  • Use as primary references or spectral sources
These long-lasting PEN-RAY lamps are rated for 5000 hours of use. Economically calibrate your photometric and fluorometric instruments. Lamps feature a double bore quartz tube—an excellent transmitter of UV radiation. Choose from standard, deluxe, or extra-long models. Order optional lamp shield for deluxe model 97606-00. All lamps have a 16" cable to plug into power supply. Lamps operate on 115 or 220 VAC—order power supply separately. Shpg wt 1.0 lb (0.5 kg).
111,000.00 - ₹255,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Bulb / Tube Life Wavelength
Availability Pricing
Analytik Jena Pen-Ray 90001201 Standard UV lamp; 254 nm, 2.13" 9760600 Mfr # 90-0012-01
COMPARE
5000 hours 254
₹121,000.00INR / Each
Analytik Jena Pen-Ray 90001901 Standard UV lamp; 365 nm, 2.25" 9760602 Mfr # 90-0019-01
COMPARE
5000 hours 365
₹111,000.00INR / Each
Analytik Jena Pen-Ray 90000401 UV Light Source, 254 nm, 9" Length 9760608 Mfr # 90000401
COMPARE
5000 hours 254
₹255,000.00INR / Each