Cole-Parmer Replacement tube for 105 x 75 mm and 200 x 40 mm UV fluorescent table, 254 nm

Representative image only.

Cole-Parmer Replacement tube for 105 x 75 mm and 200 x 40 mm UV fluorescent table, 254 nm

Cole-Parmer | Item: 0981546

₹21,600.00INR / Each

₹21,600.00INR / Each

Buy It Together

Current Item

Cole-Parmer Replacement tube for 105 x 75 mm and 200 x 40 mm UV fluorescent table, 254 nm
Cole-Parmer Replacement tube for 105 x 75 mm and 200 x 40 mm UV fluorescent table, 254 nm
0981546

₹21,600.00INR / Each
+ + + + + +
Ultraviolet Crosslinker Replacement Tube; 302 nm
9760751

₹17,100.00INR / Each
=
Total Price 391300.00 ADD 7 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Keep your ultraviolet equipment running and dependable with replacement bulbs and accessories.

Product may contain trace amounts of mercury; cautions should be used in breakage, cleanup, shipping and/or disposal.

Specifications
  • DescriptionReplacement tube for 105 x 75 mm and 200 x 40 mm UV fluorescent table, 254 nm
Reviews
Q&A