Cole-Parmer Optional cover for beaker 07206-80
Representative image only.
Cole-Parmer Optional cover for beaker 07206-80
Cole-Parmer | Item: 0720780
Reduce evaporation with beaker covers
₹11,300.00INR / Each
₹11,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer Optional cover for beaker 07206-80
Cole-Parmer Optional cover for beaker 07206-80
0720780
₹11,300.00INR / Each
+ +
Cole-Parmer Utility Tank, 304 SS, 1-1/4 gallon
0723505
₹15,100.00INR / Each
+
Cole-Parmer Stainless Steel Pouring Beaker, 2 qt
0720820
₹23,800.00INR / Each
+
Cole-Parmer Stainless Steel Lab Beaker, 1.9 L (2 Qt)
0720620
₹8,700.00INR / Each
+
Cole-Parmer Covered Container, 304 SS, 1/2 qt
0725000
₹21,500.00INR / Each
+ + =
Total Price 117700.00 ADD 8 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
These stainless steel covers help reduce evaporation, while containing harmful vapors and keeping contaminants out.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithOptional cover for beaker 07206-80
  • DescriptionAccessory Optional cover for beaker 07206-80