Product Type
In Stock
Industry
Reservoir Capacity (L)
Max Temperature (° C)
Tubes Held
Promos and Programs
Bath and Circulator Accessories
The product searched is discontinued, please browse below for replacements.
Bath and Circulator Accessories
Add bath and circulator accessories to get the most out of your baths and circulators. When filling your bath or circulator, use bath fluids to maintain desired bath temperatures, outside of the range for water. For operator safety, set the overtemperature cutoff of your bath or circulator below the flash point of the bath fluid. We offer hollow bath ball covers, lids, tanks, test tube racks, and bath fluids for your convenience.
APPLY FILTERS
filter byclear all
18 RESULTS
SORT BY
Showing 1 - 18 of 18