Lab Armor Metallic Thermal Conductivity Beads
Hold containers in place at any angle without any float-away or ruined samples
  • High thermal conductivity ±180°C
  • Versatile enough for freezers, ovens and incubators
Smooth, rounded surfaces measure 5 to 8 mm diameter and 1 to 2 mm high. Beads are moisture and gas impermeable, and are easy to keep clean with out harmful germicides. These versatile beads can also be used in a container for freezer, ovens and incubators.
55,300.00 - ₹221,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
₹55,300.00INR / Each
COMPARE
₹111,000.00INR / Each
COMPARE
₹221,000.00INR / Each