Metallic Bath Beads for Dry Baths; 8-Liters
Representative image only.
Metallic Bath Beads for Dry Baths; 8-Liters
Lab Armor| Item: 0627452
Hold containers in place at any angle without any float-away or ruined samples
₹263,000.00INR / Each
₹263,000.00INR / Each
Buy It Together
Current Item
Metallic Bath Beads for Dry Baths; 8-Liters
Metallic Bath Beads for Dry Baths; 8-Liters
0627452
₹263,000.00INR / Each
+
Beads for dry bath, 0.75-liter (Beads and Chill Packs)
0627438
₹24,500.00INR / Each
+ =
Total Price 655500.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • High thermal conductivity ±180°C
  • Versatile enough for freezers, ovens and incubators
  • 1 year warranty
More About this Item
Smooth, rounded surfaces measure 5 to 8 mm diameter and 1 to 2 mm high. Beads are moisture and gas impermeable, and are easy to keep clean with out harmful germicides. These versatile beads can also be used in a container for freezer, ovens and incubators.