Beads for dry bath, 0.75-liter (Beads and Chill Packs)
Representative image only.
Beads for dry bath, 0.75-liter (Beads and Chill Packs)
Lab Armor| Item: 0627438
Replace water in laboratory water baths and ice in ice buckets.
₹24,500.00INR / Each
₹24,500.00INR / Each
Buy It Together
Current Item
Beads for dry bath, 0.75-liter (Beads and Chill Packs)
Beads for dry bath, 0.75-liter (Beads and Chill Packs)
0627438
₹24,500.00INR / Each
+
Metallic Bath Beads for Dry Baths; 2-Liters
0627450
₹83,000.00INR / Each
=
Total Price 107500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Reduce the risk of contamination, achieve reproducible results and have greater confidence in your work
  • No need to worry about burnout as there is no water to evaporate.
  • Highly recyclable, fewer harmful biocides, and more energy efficient beads are eco-friendly
  • Constructed of durable metal with high thermal conductivity
  • Moisture and gas impermeable with a smooth, round surface
  • Working temperature range is -80°C to 180°C
  • 1 year warranty
More About this Item
Compatible with waterless ice buckets, items 06274-24 and -28.
These beads are eco-friendly and mproves energy efficiency of standard water baths by over 50%.
1 Year Warranty
Specifications
  • DescriptionBeads for dry bath, 0.75-liter (Beads and Chill Packs)