Accessories Cole-Parmer® CB-200D-IB Electronic Ice Bucket
Keep your samples cold, clean, and dry
These Techne N°ICE ceramic-coated beads maintain the temperature and support the sample vessels for as long as necessary. The beads are chemically resistant and if required can be autoclaved at 134°C. As a time saving option, the bucket of beads can be incubated in the fridge overnight and simply inserted into the unit as and when required. Compatible with the 24120-00 Electronic Ice Bucket, sold separately.
12,700.00 - ₹17,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Electronic Ice Bucket Ceramic Beads
₹12,700.00INR / Each
COMPARE
Electronic Ice Bucket Spare Bucket
₹17,900.00INR / Each