Temperature Bath Fluids
Get more out of your bath!
  • Maintains bath temperature as high as 280°C
  • 0ver temperature cutoff provides added operator safety
Use bath fluids to maintain desired bath temperatures, outside of the range for water. For operator safety, set the overtemperature cutoff of your bath/circulator below the flash point of the bath fluid.
Note: We do not recommend using automotive anti-freeze with circulating baths — additives can be harmful to the circulating pump. Silicone bath fluids are not compatible with silicone tubing.
72,200.00INR / Gallon of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
PolyScience Dynalene Temperature Bath Fluid, -50 to 100 °C; 1 Gal (3.8 L) 0129700 Mfr # 060330
COMPARE
100 -50 Temperature Bath Fluid, -50 to 100 °C; 1 Gal (3.8 L)
₹72,200.00INR / Gallon of 1