Cole-Parmer Disposable Bottle Top Aspirator System, SFCA Membrane, 0.45 um, 250mL, Sterile; 12/CS

Representative image only.

Cole-Parmer Disposable Bottle Top Aspirator System, SFCA Membrane, 0.45 um, 250mL, Sterile; 12/CS

Cole-Parmer | Item: 2440669

Save time with presterilized disposable filtration units

₹21,600.00INR / Case of 12

₹21,600.00INR / Case of 12

Buy It Together

Current Item

Cole-Parmer Disposable Bottle Top Aspirator System, SFCA Membrane, 0.45 um, 250mL, Sterile; 12/CS
Cole-Parmer Disposable Bottle Top Aspirator System, SFCA Membrane, 0.45 um, 250mL, Sterile; 12/CS
2440669

₹21,600.00INR / Case of 12
+ + + + =
Total Price 325800.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Individually wrapped for sterility
  • Easy-pouring large cap on reservoir
  • Graduations on funnel and reservoir for improved accuracy
  • Hose connector fits multiple hose diameters

More About this Item

Disposable bottletop aspirators provide a complete vacuum filtration solution for your separation and purification needs. Intended for the sterile vacuum filtration of aqueous solutions such as cell culture media and biological fluids.

All units come presterilized by gamma irradiation and certified nonpyrogenic, and are individually wrapped in easy-to-open packages. The large openings on the reservoir and funnel make for easy access. The housing (cup, bottle, and lid) are made of clear polystyrene for easy viewing. The filter tip is constructed of polypropylene, while the joint connector is made of Acrylonitrile Butadiene Styrene (ABS) and the cap material is High-Density Polyethylene (HDPE).

Surfactant-free cellulose acetate filter membranes contain none of the wetting agents found in standard CA (cellulose acetate). These wetting agents have been shown to be toxic to specific cell lines in vitro (1-3). SFCA also has lower protein binding than does standard CA.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

Specifications
  • Membrane MaterialSurfactant-free Cellulose Acetate (SFCA)
  • Pore Size (µm)0.45
  • Membrane Diameter (mm)50
  • Housing MaterialPolystyrene (PS)
  • Capacity (mL)250
  • SterileYes
  • ConnectionsHose Barb
  • ColorClear
  • DescriptionDisposable Bottle Top Aspirator, SFCA Membrane, 0.45um, 250mL, Sterile; 12/cs
Reviews
Q&A