Cole-Parmer® Disposable Bottletop Aspirators
Save time with presterilized disposable filtration units
  • Individually wrapped for sterility
  • Easy-pouring large cap on reservoir
  • Graduations on funnel and reservoir for improved accuracy
  • Hose connector fits multiple hose diameters
Disposable bottletop aspirators provide a complete vacuum filtration solution for your separation and purification needs. Intended for the sterile vacuum filtration of aqueous solutions such as cell culture media and biological fluids.
All units come presterilized by gamma irradiation and certified nonpyrogenic, and are individually wrapped in easy-to-open packages. The large openings on the reservoir and funnel make for easy access. The housing (cup, bottle, and lid) are made of clear polystyrene for easy viewing. The filter tip is constructed of polypropylene, while the joint connector is made of Acrylonitrile Butadiene Styrene (ABS) and the cap material is High-Density Polyethylene (HDPE).
10,700.00 - ₹44,900.00INR / Pkg of 12
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
150 Polyethersulfone (PES) 0.22
₹11,000.00INR / Pkg of 12
COMPARE
250 Polyethersulfone (PES) 0.22
₹12,400.00INR / Pkg of 12
COMPARE
500 Polyethersulfone (PES) 0.22
₹15,300.00INR / Pkg of 12
COMPARE
1000 Polyethersulfone (PES) 0.22
₹24,900.00INR / Pkg of 12
COMPARE
250 Polyvinylidene fluoride (PVDF) 0.22
₹12,400.00INR / Pkg of 12
COMPARE
250 Polyvinylidene fluoride (PVDF) 0.45
₹12,400.00INR / Pkg of 12
COMPARE
500 Polyvinylidene fluoride (PVDF) 0.22
₹15,300.00INR / Pkg of 12
COMPARE
500 Polyvinylidene fluoride (PVDF) 0.45
₹15,300.00INR / Pkg of 12
COMPARE
1000 Polyvinylidene fluoride (PVDF) 0.22
₹24,900.00INR / Pkg of 12
COMPARE
1000 Polyvinylidene fluoride (PVDF) 0.45
₹24,900.00INR / Pkg of 12
COMPARE
250 Nylon 0.22
₹10,700.00INR / Pkg of 12
COMPARE
250 Nylon 0.45
₹12,400.00INR / Pkg of 12
COMPARE
500 Nylon 0.22
₹13,200.00INR / Pkg of 12
COMPARE
500 Nylon 0.45
₹15,300.00INR / Pkg of 12
COMPARE
500 Polyethersulfone (PES) 0.22
₹23,200.00INR / Pkg of 24
COMPARE
250 Surfactant-Free Cellulose Acetate (SFCA) 0.22
₹22,500.00INR / Case of 12
COMPARE
250 Surfactant-free Cellulose Acetate (SFCA) 0.45
₹21,600.00INR / Case of 12
COMPARE
500 Surfactant-Free Cellulose Acetate (SFCA) 0.22
₹26,600.00INR / Case of 12
COMPARE
500 Surfactant-free Cellulose Acetate (SFCA) 0.45
₹26,600.00INR / Case of 12
COMPARE
1000 Surfactant-Free Cellulose Acetate (SFCA) 0.22
₹44,900.00INR / Case of 12
Showing 1 - 20 of 21