Environmental Express Assembled Filter Funnel, PP and PTFE, 0.2 mu, 50/pk
Representative image only.
Environmental Express Assembled Filter Funnel, PP and PTFE, 0.2 mu, 50/pk
Environmental Express | Item: 3520036
Convenient units are perfect for vacuum filtration
₹17,600.00INR / Pkg of 1
₹17,600.00INR / Pkg of 1
Buy It Together
Current Item
Environmental Express Assembled Filter Funnel, PP and PTFE, 0.2 mu, 50/pk
Environmental Express Assembled Filter Funnel, PP and PTFE, 0.2 mu, 50/pk
3520036
₹17,600.00INR / Pkg of 1
+ + =
Total Price 52200.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for larger volume filtration
  • Comes standard with pre-inserted filters
More About this Item
These filters fit snugly in most standard vacuum flasks. There are preassembled and disposable options perfect for any number of applications.
Assembled filter funnels with filters include a top cap, filter support and filter.
Specifications
  • Membrane MaterialPolytetrafluoroethylene (PTFE)
  • Pore Size (µm)0.2
  • Membrane Diameter (mm)25
  • Housing MaterialPolypropylene (PP)
  • Sample Volume (mL)68
  • DescriptionAssembled Filter Funnel, PP and PTFE, 0.2 mu, 50/pk