Thermo Scientific Nalgene Disposable Analytical Filter Units
Cellulose nitrate (CN) membrane provides for the superior growth of microorganisms and is triton free
  • Upper twists off easily for easy access to 47-mm dia membrane
These Nalgene® filter units are designed for microbiological analysis testing of water, food and beverage, raw materials, and finished products. Units with 0.45 µm membranes meet requirements for APHA Standard Methods and U.S. EPA water quality testing. All units come presterilized.
ilter Units are comprised of polypropylene upper and high-impact polystyrene lower. Units include a 1/4" to 3/8" ID quick-disconnect tubing adapter for vacuum connection. Order optional filter unit holder under Accessories.
42,000.00 - ₹42,600.00INR / Dozen of 12
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Pore Size (µm)
Availability Pricing
Thermo Scientific Nalgene 130-4020 Analytical Filter Units, 0.2um, 150 mL; 12/Pk 0673002 Mfr # 130-4020
COMPARE
150 0.2
₹42,600.00INR / Dozen of 12
Thermo Scientific Nalgene 130-4045 Analytical Filter Units, 0.45um, 150 mL; 12/Pk 0673004 Mfr # 130-4045
COMPARE
150 0.45
₹42,000.00INR / Dozen of 12