Cole-Parmer Blue Light Transilluminator Replacement Amber Cover; 1/Ea

Representative image only.

Cole-Parmer Blue Light Transilluminator Replacement Amber Cover; 1/Ea

Analytik Jena | Item: 9762382

₹12,700.00INR / Each

₹12,700.00INR / Each

Buy It Together

Current Item

Cole-Parmer Blue Light Transilluminator Replacement Amber Cover; 1/Ea
Cole-Parmer Blue Light Transilluminator Replacement Amber Cover; 1/Ea
9762382

₹12,700.00INR / Each
+ + + + +
Ultraviolet Crosslinker Replacement Tube; 302 nm
9760751

₹17,100.00INR / Each
+ +
Cole-Parmer Blue Light Transilluminator; 115V
9762381

₹188,000.00INR / Each
=
Total Price 570400.00 ADD 8 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Keep your ultraviolet equipment running and dependable with replacement bulbs and accessories.

Specifications
  • Compatible With97623-81 or 97623-83
  • DescriptionBlue Light Transilluminator Replacement Amber Cover; 1/Ea
Reviews
Q&A