Cole-Parmer 9/21 Degree Baume Shatterproof Plastic Hydrometer
Representative image only.
Cole-Parmer 9/21 Degree Baume Shatterproof Plastic Hydrometer
Cole-Parmer | Item: 0829828
₹35,100.00INR / Each
for an additional
₹27,300.00INR / Each
for an additional
₹39,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹35,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer 9/21 Degree Baume Shatterproof Plastic Hydrometer
Cole-Parmer 9/21 Degree Baume Shatterproof Plastic Hydrometer
0829828
₹35,100.00INR / Each
+ + + + +
Cole-Parmer 0/12 Degree Baume Hydrometer
0829944
₹9,300.00INR / Each
+ +
InnoCal NIST-Traceable Calibration; Hydrometer, 1-point
1710700
₹27,300.00INR / Each
+ + =
Total Price 142500.00 ADD 10 ITEMS TO CART
Product Description
Includes
Choose from our comprehensive line of Hydrometers and other Hydrometers products.
Key Features
  • 5 year warranty
More About this Item
These durable polycarbonate hydrometers won''t break when dropped—perfect for industrial or laboratory use.
The temperature standard for these hydrometers is 60°F (15°C). All hydrometers are individually serialized, feature metal ballast with wax epoxy and include multi-language statement of accuracy supplied by the vendor.
These hydrometers are available in models measuring Baume of syrups and liquids heavier than water in various ranges from 0 to 31°.
Note: Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.
Equivalent to H-B Instrument 618000800
5 Year Warranty
Specifications
  • Length (")10
  • Hydrometer FormPlain
  • Baume Range (Max) (° C)21
  • Baume Range (Min) (° C)9
  • Division0.2°
  • Accuracy±0.2°
  • Length (mm)255
  • Description 9/21 Degree Baume Shatterproof Plastic Hydrometer
  • Warranty5 years