Cole-Parmer Baume Plain Form Hydrometers
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
For liquids heavier than water, each hydrometer is calibrated in our triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
6,200.00 - ₹6,800.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0/12 Degree Baume Hydrometer
₹6,500.00INR / Each
COMPARE
9/21 Degree Baume Hydrometer
₹6,500.00INR / Each
COMPARE
19/31 Degree Baume Hydrometer
₹6,500.00INR / Each
COMPARE
29/41 Degree Baume Hydrometer
₹6,500.00INR / Each
COMPARE
39/51 Degree Baume Hydrometer
₹6,500.00INR / Each
COMPARE
49/61 Degree Baume Hydrometer
₹6,800.00INR / Each
COMPARE
59/71 Degree Baume Hydrometer
₹6,800.00INR / Each
COMPARE
0/35 Degree Baume Hydrometer
₹6,400.00INR / Each
COMPARE
35/70 Degree Baume Hydrometer
₹6,500.00INR / Each
COMPARE
0/50 Degree Baume Hydrometer
₹6,200.00INR / Each
COMPARE
0/70 Degree Baume Hydrometer
₹6,400.00INR / Each
Showing 1- 11 of 11