Cole-Parmer Polycarbonate Baume Hydrometers, plain form
These durable polycarbonate hydrometers won''t break when dropped—perfect for industrial or laboratory use.
The temperature standard for these hydrometers is 60°F (15°C). All hydrometers are individually serialized, feature metal ballast with wax epoxy and include multi-language statement of accuracy supplied by the vendor.
These hydrometers are available in models measuring Baume of syrups and liquids heavier than water in various ranges from 0 to 31°.
Note: Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.
24,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0/12 Degree Baume Shatterproof Plastic Hydrometer
₹24,400.00INR / Each
COMPARE
9/21 Degree Baume Shatterproof Plastic Hydrometer
₹24,400.00INR / Each
COMPARE
19/31 Degree Baume Shatterproof Plastic Hydrometer
₹24,400.00INR / Each