Chemetrics R-9010 Refill Kit for 05541-04, 30 Tests
Representative image only.
Chemetrics R-9010 Refill Kit for 05541-04, 30 Tests
Chemetrics | Mfr: R-9010| Item: 0554107
Take it wherever you go
₹12,700.00INR / Each
₹12,700.00INR / Each
Buy It Together
Current Item
Chemetrics R-9010 Refill Kit for 05541-04, 30 Tests
Chemetrics R-9010 Refill Kit for 05541-04, 30 Tests
0554107
₹12,700.00INR / Each
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
+ + =
Total Price 55900.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed packaging keeps reagents active
  • 2 year warranty
More About this Item
CHEMets colorimetric test kits let you visually compare the color of your sample to standards using a comparator, comparator sold separately. Just break tip of ampoule in cup filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator provides accurate visual analysis.
Two year warranty
Specifications
  • Compatible With05541-04
  • DescriptionRefill Kit for 05541-04, 30 Tests