Chemetrics R-8012 Refill Kit , PHenol Test, for 00290-30
Representative image only.
Chemetrics R-8012 Refill Kit , PHenol Test, for 00290-30
Chemetrics | Mfr: R-8012| Item: 0029032
Take it wherever you go
₹14,300.00INR / Box of 30
₹14,300.00INR / Box of 30
Buy It Together
Current Item
Chemetrics R-8012 Refill Kit , PHenol Test, for 00290-30
Chemetrics R-8012 Refill Kit , PHenol Test, for 00290-30
0029032
₹14,300.00INR / Box of 30
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
+ + =
Total Price 58700.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed packaging keeps reagents active
More About this Item
CHEMets colorimetric test kits let you visually compare the color of your sample to standards using a comparator, comparator sold separately. Just break tip of ampoule in cup filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator provides accurate visual analysis.
Specifications
  • Compatible With00290-30
  • DescriptionRefill Kit , PHenol Test, for 00290-30