Chemetrics R-6001 Refill Kit Iron, Total Soluble Test, for 00291-40
Representative image only.
Chemetrics R-6001 Refill Kit Iron, Total Soluble Test, for 00291-40
Chemetrics | Mfr: R-6001| Item: 0029141
Take it wherever you go
Call For Price
Call For Price
Buy It Together
Current Item
Chemetrics R-6001 Refill Kit Iron, Total Soluble Test, for 00291-40
Chemetrics R-6001 Refill Kit Iron, Total Soluble Test, for 00291-40
0029141
Call For Price
+
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050
Call For Price
+ + =
Total Price 43200.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Vacuum sealed packaging keeps reagents active
More About this Item
CHEMets colorimetric test kits let you visually compare the color of your sample to standards using a comparator, comparator sold separately. Just break tip of ampoule in cup filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator provides accurate visual analysis.
Not Available for Canada.
Specifications
  • Compatible With00291-40
  • DescriptionRefill Kit Iron, Total Soluble Test, for 00291-40