Bullard EVAHL20TICT EVA PAPR Explosion Proof System with CC20 Hood; 1 Set

Representative image only.

Bullard EVAHL20TICT EVA PAPR Explosion Proof System with CC20 Hood; 1 Set

Bullard | Mfr: EVAHL20TICT | Item: 1635916

Combining the cutting-edge technology and ergonomic design of the EVA, with approval for use in Hazardous Locations

₹256,000.00INR / Set of 1

₹256,000.00INR / Set of 1

Buy It Together

Current Item

Bullard EVAHL20TICT EVA PAPR Explosion Proof System with CC20 Hood; 1 Set
Bullard EVAHL20TICT EVA PAPR Explosion Proof System with CC20 Hood; 1 Set
1635916

₹256,000.00INR / Set of 1
+ =
Total Price 312100.00 ADD 2 ITEMS TO CART
Product Description

Includes

PAPR Assemblies include blower (EVA1), belt (EVABELT1), battery (EVAHLBAT1), breathing tube(PAHBT), charger (EVASMC), airflow indicator (PA1AFI), and double-bib hood; filter is not included.

Key Features

  • Due to the high demand for personal safety products, cancellations and returns will be restricted.

  • 1 year warranty

More About this Item


HazLoc Rated:
• For Class I, Division 2, Groups A, B, C, D T6 (Gases and Vapors)
• Class II, Division 2, Groups F, G (Dusts)
• Class III (Fibers or Flyings)

Simple:

• One-Button Operation
• Choice of Two Air Speeds
• Single-Filter Design

Safe:

• Active Flow Technology Responds to AirFlow Conditions
• 1,000 Assigned Protection Factor (APF) Independent, 3rd Party Documentation
• Dual Alarms for Low Flow and Low Battery

Dependable:

• Visual Fuel Gauge
• 10X the Life of Most Brush Motors
• Genius Charger Compatible

Combining the cutting-edge technology and ergonomic design of the EVA, with approval for use in Hazardous Locations, the EVAHL Powered Air Purifying Respirator (PAPR) offers users the latest in respiratory protection for explosive environments.

• Class I, Division 2, Groups A, B, C, D, T6 (Gases and Vapors)
• Class II, Division 2 Groups F, G (Dusts)
• Class III (Fibers or Flyings)
• CSA International Certificate 2510928 per ANSI/ISA 2.12.01-2007

Equipped with an innovative Active Flow Technology this intelligent system is continuously working to maintain constant airflow, regardless of filter type, hood type, filter loading or battery capacity. Plus, EVA is equipped with an easy-to-read fuel gauge that lets the user quickly check the unit’s battery status.

Applications:
• Healthcare
• Pharmaceutical Manufacturing
• Paint Respirator and Coating Respirator
• Welding Respirator and Grinding Respirator
• Laboratory
• Animal Science

1 Year Warranty
Specifications
  • ComplianceNIOSH
  • DescriptionBullard® EVAHL20TICT EVA PAPR Explosion Proof System with CC20 Hood; 1 Set
  • Warranty1 year
Reviews
Q&A