Bullard® EVAHL Powered Air-Purifying Respirator System

Combining the cutting-edge technology and ergonomic design of the EVA, with approval for use in Hazardous Locations
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.


HazLoc Rated:
• For Class I, Division 2, Groups A, B, C, D T6 (Gases and Vapors)
• Class II, Division 2, Groups F, G (Dusts)
• Class III (Fibers or Flyings)

Simple:

• One-Button Operation
• Choice of Two Air Speeds
• Single-Filter Design

Safe:

• Active Flow Technology Responds to AirFlow Conditions
• 1,000 Assigned Protection Factor (APF) Independent, 3rd Party Documentation
• Dual Alarms for Low Flow and Low Battery

Dependable:

• Visual Fuel Gauge
• 10X the Life of Most Brush Motors
• Genius Charger Compatible

Combining the cutting-edge technology and ergonomic design of the EVA, with approval for use in Hazardous Locations, the EVAHL Powered Air Purifying Respirator (PAPR) offers users the latest in respiratory protection for explosive environments.

• Class I, Division 2, Groups A, B, C, D, T6 (Gases and Vapors)
• Class II, Division 2 Groups F, G (Dusts)
• Class III (Fibers or Flyings)
• CSA International Certificate 2510928 per ANSI/ISA 2.12.01-2007

Equipped with an innovative Active Flow Technology this intelligent system is continuously working to maintain constant airflow, regardless of filter type, hood type, filter loading or battery capacity. Plus, EVA is equipped with an easy-to-read fuel gauge that lets the user quickly check the unit’s battery status.

Applications:
• Healthcare
• Pharmaceutical Manufacturing
• Paint Respirator and Coating Respirator
• Welding Respirator and Grinding Respirator
• Laboratory
• Animal Science
256,000.00INR / Set of 1

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Bullard EVAHL20TICT EVA PAPR Explosion Proof System with CC20 Hood; 1 Set 1635916 Mfr # EVAHL20TICT
COMPARE
Bullard® EVAHL20TICT EVA PAPR Explosion Proof System with CC20 Hood; 1 Set
₹256,000.00INR / Set of 1
Bullard EVAHLRT3 EVA PAPR Explosion Proof System with RT3 Hood; 1 Set 1635918 Mfr # EVAHLRT3
COMPARE
Bullard® EVAHLRT3 EVA PAPR Explosion Proof System with RT3 Hood; 1 Set
₹256,000.00INR / Set of 1
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat