Bullard RT3/RT3T Tychem® QC Hoods for EVA Respirator without ratchet suspension, Wide Angle Lens; 10/Pk
Representative image only.
Bullard RT3/RT3T Tychem® QC Hoods for EVA Respirator without ratchet suspension, Wide Angle Lens; 10/Pk
Bullard | Mfr: RT3/RT3T| Item: 1635924
Provide a wide variety of simple, effective and economical options for on-the-job respiratory protection.
₹101,000.00INR / Pkg of 10
₹101,000.00INR / Pkg of 10
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Available in multiple variants
More About this Item
1. CC20 Series DuPont™ Tychem® Series Hood:
The CC20 Series of DuPont™ Tychem® 2000 Hoods provide a wide variety of simple, effective and economical options for on-the-job respiratory protection.
A proprietary neck cuff design helps maintain positive pressure inside the hood to aid in keeping contaminants out. As air enters the hood, the inner collar inflates, providing a snug, comfortable fit around the neck and continuous flow of air to the worker’s breathing zone. Racheted for user comfort covering the entire head, face and neck, the CC20 hood provides splash/overspray protection and makes cleanup quick and easy.
Key Features:
• Approved by NIOSH for use in Type C applications
• OSHA Assigned Protection Factor (APF) of 1,000
• Tested against numerous chemical warfare agents and found effective for limited use. See 2003 Permeation Guide
• CC20 hood is lighter and more comfortable than tight-fitting full-facepiece respirator
• Covering the entire head, face, and neck, hoods provide splash/overspray protection and makes cleanup quick and easy
• One size fits all
• Double Bib (Inner Bib) Style: Outer bib with a long inner bib; tuck inner bib into shirt or coveralls to funnel excess air from the hood to the body
Applications:
Include Pharmaceutical manufacturing, Chemical and Pesticide handling, Painting and coating operation, and Nuclear operation
2. RT Series DuPont™ Tychem® Hoods:
The headband-free RT Series hood provides high performance respiratory protection through a comfortable sport neck cuff design.
Comfortable sport neck cuff design that helps maintain positive pressure inside the hood, helping to keep contaminants out. The patented proprietary over-the-top air delivery system delivers the air over the user’s head, down the front of the lens, and into the breathing zone.
Key Features:
• Headband-free design provides better comfort
• 1,000 Assigned Protection Factor (APF)
• Over-the-Top Airflow Delivery System reduces possibility of dry eyes and nasal passages, provides cooling airflow to the torso and promotes curvature of the hood for a better fit.
• Panoramic Viewing Lens provides 135 square inches of unobstructed visibility
• Sport Neck Cuff Designed to provide soft, comfortable protection around the user’s neck, making head/neck movement easy, reducing any rubbing.
• No Fit Testing, One size fits all
• Long, Double Bibs. The longer bibs help users keep hoods tucked in when moving the arms and shoulders, while also allowing excess air to travel to the torso for a cooling effect.
Conformite Europeenne
Specifications
  • DescriptionBullard® RT3/RT3T Tychem® QC Hoods for EVA Respirator without ratchet suspension, wide angle lens; 10/Pk