Analytik Jena Pen-Ray 99005502 Power supply for 97606-00 and -02, 220 VAC/50 Hz

Representative image only.

Analytik Jena Pen-Ray 99005502 Power supply for 97606-00 and -02, 220 VAC/50 Hz

Cole-Parmer | Mfr: 99-0055-02| Item: 9760685

₹123,000.00INR / Each

₹123,000.00INR / Each

Buy It Together

Current Item

Analytik Jena Pen-Ray 99005502 Power supply for 97606-00 and -02, 220 VAC/50 Hz
Analytik Jena Pen-Ray 99005502 Power supply for 97606-00 and -02, 220 VAC/50 Hz
9760685

₹123,000.00INR / Each
+ + + + +
Ultraviolet Crosslinker Replacement Tube; 302 nm
9760751

₹17,100.00INR / Each
+ =
Total Price 492700.00 ADD 7 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Keep your ultraviolet equipment running and dependable with replacement bulbs and accessories.

1 Year Warranty
Specifications
  • Compatible With97606-00 and 97606-02
  • Power (VAC)220
  • DescriptionPower supply for 97606-00 and -02, 220 VAC/50 Hz
  • Warranty1 year
Reviews
Q&A