Analytik Jena Pen-Ray 98000802 Lamp shield, 5/16" x 5/8" opening

Representative image only.

Analytik Jena Pen-Ray 98000802 Lamp shield, 5/16" x 5/8" opening

Cole-Parmer | Mfr: 98-0008-02| Item: 9760652

₹18,500.00INR / Each

₹18,500.00INR / Each

Buy It Together

Current Item

Analytik Jena Pen-Ray 98000802 Lamp shield, 5/16" x 5/8" opening
Analytik Jena Pen-Ray 98000802 Lamp shield, 5/16" x 5/8" opening
9760652

₹18,500.00INR / Each
+ + + + +
Ultraviolet Crosslinker Replacement Tube; 302 nm
9760751

₹17,100.00INR / Each
+ =
Total Price 388200.00 ADD 7 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Keep your ultraviolet equipment running and dependable with replacement bulbs and accessories.

This accessory is available for deluxe model 97606-00.

Use to direct UV light through a point-like aperture or small window opening. Shield also protects tube from breakage.

1 Year Warranty
Specifications
  • Compatible With97606-00
  • DescriptionLamp shield, 5/16" x 5/8" opening
  • Warranty1 year
Reviews
Q&A