VELP KjTabs™ Catalyst Tablets
Speed up digestions
  • Compatible with any digestion system Wide variety of catalyst formulae work with most samples
Use with the Velp DK and DKL Digestion Systems, or any other similar system.
36,200.00 - ₹118,000.00INR / Box of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Materials of Construction Description
Availability Pricing
Velp KjTabs Catalyst Tablets, VS Antifoam; 1000/BX WW-86579-97 Mfr # A00000283
COMPARE
0.97 g Na2SO4 + 0.03 g Silicone (antifoam) Catalyst Tablets, VS Antifoam; 1000/BX
₹90,100.00INR / Box of 1000
Velp KjTabs Catalyst Tablets, VCT; 1000/BX WW-86580-05 Mfr # A00000276
COMPARE
5 g K2SO4 + 0.15 g CuSO4 x 5 H2O + 0.15 g TiO2 Catalyst Tablets, VCT; 1000/BX
₹118,000.00INR / Box of 1000
Velp KjTabs Catalyst Tablets, VCM; 1000/BX WW-86580-08 Mfr # A00000274
COMPARE
3.5 g K2SO4 + 0.1 g CuSO4 x 5 H2O Catalyst Tablets, VCM; 1000/BX
₹36,200.00INR / Box of 1000