Velp KjTabs Catalyst Tablets, VCT; 1000/BX
Representative image only.
Velp KjTabs Catalyst Tablets, VCT; 1000/BX
Velp | Mfr: A00000276| Item: 8658005
Speed up digestions
₹148,000.00INR / Box of 1000
₹148,000.00INR / Box of 1000
Product Description
Key Features
  • Compatible with any digestion system Wide variety of catalyst formulae work with most samples
  • Formulation: 5 g K2SO4 + 0.15 g CuSO4 x 5 H2O + 0.15 g TiO2
  • 2 year warranty
More About this Item
Use with the Velp DK and DKL Digestion Systems, or any other similar system.
Two year warranty
Specifications
  • Materials of Construction5 g K2SO4 + 0.15 g CuSO4 x 5 H2O + 0.15 g TiO2
  • DescriptionCatalyst Tablets, VCT; 1000/BX