Velp KjTabs Catalyst Tablets, VS Antifoam; 1000/BX
Representative image only.
Velp KjTabs Catalyst Tablets, VS Antifoam; 1000/BX
Velp | Mfr: A00000283| Item: 8657997
Speed up digestions
₹116,000.00INR / Box of 1000
₹116,000.00INR / Box of 1000
Product Description
Key Features
  • Compatible with any digestion system Wide variety of catalyst formulae work with most samples
  • Formulation: 0.97 g Na2SO4 + 0.03 g Silicone (antifoam)
  • 2 year warranty
More About this Item
Use with the Velp DK and DKL Digestion Systems, or any other similar system.
Two year warranty
Specifications
  • Materials of Construction0.97 g Na2SO4 + 0.03 g Silicone (antifoam)
  • DescriptionCatalyst Tablets, VS Antifoam; 1000/BX