Velp KjTabs Catalyst Tablets, VCM; 1000/BX
Representative image only.
Velp KjTabs Catalyst Tablets, VCM; 1000/BX
Velp | Mfr: A00000274| Item: 8658008
Speed up digestions
₹65,830.00INR / Box of 1000
₹65,830.00INR / Box of 1000
Product Description
Key Features
  • Compatible with any digestion system Wide variety of catalyst formulae work with most samples
  • Formulation: 3.5 g K2SO4 + 0.1 g CuSO4 x 5 H2O
  • 2 year warranty
More About this Item
Use with the Velp DK and DKL Digestion Systems, or any other similar system.
Two year warranty
Specifications
  • Materials of Construction3.5 g K2SO4 + 0.1 g CuSO4 x 5 H2O
  • DescriptionCatalyst Tablets, VCM; 1000/BX