Trajan SilTite Fittings
Available in two material types: stainless steel and Glass Lined Tubing
  • Stainless steel unions come in a variety of tube sizes and configurations
  • GL T unions are inert and mainly used for connecting fused silica tubing of various internal diameters
9,800.00 - ₹22,200.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 073550 SilTite™ Stainless Steel Mini Union, Compatible with 0.4 mm ID Columns; 1/Pk 1600851 Mfr # 073550
COMPARE
₹13,100.00INR / Each
Trajan SGE 073551 SilTite™ Stainless Steel Mini Union, for 0.5 mm ID Columns; 1/Pk 1600852 Mfr # 073551
COMPARE
₹13,100.00INR / Each
Trajan SGE 073553 SilTite™ Nuts, Compatible with Mini Unions of all ID; 5/Pk 1600854 Mfr # 073553
COMPARE
₹9,800.00INR / Pkg of 5
Trajan SGE 073554 SilTite™ Mini Union with Ferrules, for 1/32" Columns, 0.8 mm ID; 1/Ea 1600855 Mfr # 073554
COMPARE
₹13,100.00INR / Each
Trajan SGE 073560 SilTite™ Micro Union, for 0.1-0.25 mm to 0.1-0.25 mm ID Columns/Fused Silica; 1/Ea 1600856 Mfr # 073560
COMPARE
₹22,200.00INR / Each
Trajan SGE 073561 SilTite™ Micro Union, Compatible with 0.1 - 0.25 mm and 0.32 mm Columns/Fused Silica; 1/Ea 1600857 Mfr # 073561
COMPARE
₹22,200.00INR / Each
Trajan SGE 073562 SilTite™ Micro Union, for 0.1 - 0.25 mm and 0.53 mm Columns/Fused Silica; 1/Ea 1600858 Mfr # 073562
COMPARE
₹22,200.00INR / Each
Trajan SGE 073564 SilTite™ Micro Union, Compatible with 0.32 mm and 0.53 mm Columns/Fused Silica; 1/Ea 1600860 Mfr # 073564
COMPARE
₹22,200.00INR / Each
Trajan SGE 073572 SilTite™ Micro Union, for 0.32 mm and 0.32 mm Columns/Fused Silica; 5/Pk 1600868 Mfr # 073572
COMPARE
₹20,900.00INR / Pkg of 5
Trajan SGE 073573 SilTite™ Micro Union, for 0.32 mm and 0.53 mm Columns/Fused Silica; 5/Pk 1600869 Mfr # 073573
COMPARE
₹20,900.00INR / Pkg of 5
Showing 1- 10 of 10