Trajan SGE 073573 SilTite™ Micro Union, for 0.32 mm and 0.53 mm Columns/Fused Silica; 5/Pk
Representative image only.
Trajan SGE 073573 SilTite™ Micro Union, for 0.32 mm and 0.53 mm Columns/Fused Silica; 5/Pk
Trajan | Mfr: 073573| Item: 1600869
Available in two material types: stainless steel and Glass Lined Tubing
₹20,900.00INR / Pkg of 5
₹20,900.00INR / Pkg of 5
Product Description
Includes
Supplied with the appropriate nuts, ferrules and/or sealing rings
Key Features
  • Stainless steel unions come in a variety of tube sizes and configurations
  • GL T unions are inert and mainly used for connecting fused silica tubing of various internal diameters
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE SilTite® Micro Union, for 0.32 mm and 0.53 mm Columns/Fused Silica; 5/Pk