Trajan SGE 073564 SilTite™ Micro Union, Compatible with 0.32 mm and 0.53 mm Columns/Fused Silica; 1/Ea
Representative image only.
Trajan SGE 073564 SilTite™ Micro Union, Compatible with 0.32 mm and 0.53 mm Columns/Fused Silica; 1/Ea
Trajan | Mfr: 073564| Item: 1600860
Available in two material types: stainless steel and Glass Lined Tubing
₹22,200.00INR / Each
₹22,200.00INR / Each
Product Description
Includes
Supplied with the appropriate nuts, ferrules and/or sealing rings
Key Features
  • Stainless steel unions come in a variety of tube sizes and configurations
  • GL T unions are inert and mainly used for connecting fused silica tubing of various internal diameters
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE SilTite® Micro Union, Compatible with 0.32 mm and 0.53 mm Columns/Fused Silica; 1/Ea