Trajan SGE 073553 SilTite™ Nuts, Compatible with Mini Unions of all ID; 5/Pk
Representative image only.
Trajan SGE 073553 SilTite™ Nuts, Compatible with Mini Unions of all ID; 5/Pk
Trajan | Mfr: 073553| Item: 1600854
Available in two material types: stainless steel and Glass Lined Tubing
₹9,800.00INR / Pkg of 5
₹9,800.00INR / Pkg of 5
Product Description
Includes
Supplied with the appropriate nuts, ferrules and/or sealing rings
Key Features
  • Stainless steel unions come in a variety of tube sizes and configurations
  • GL T unions are inert and mainly used for connecting fused silica tubing of various internal diameters
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE SilTite® Nuts, Compatible with Mini Unions of all ID; 5/Pk