ThermoSafe Brands Medium-Capacity Insulated Shippers

Ship nonhazardous materials
  • Add as many ice packs as you need — choose from a variety of sizes to meet your refrigeration needs.
Medium-capacity models have a rectangular interior and include cartons. Mailer 03726-53 has thick walls — save money on refrigerant.
Note: For shipping nonhazardous materials only.
4,000.00 - ₹5,400.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Inside Height (in) Inside Length (in) Inside Width (in)
Availability Pricing
ThermoSafe 318 Medium-Capacity Insulated Shipper, 8 x 6 x 4.25 in; 1/Pk 0372611 Mfr # 318
COMPARE
4 1/4 8 6
₹4,000.00INR / Each
ThermoSafe 321UPS Medium-Capacity Insulated Shipper, 8 x 5.8 x 8.8 in; 1/Pk 0372621 Mfr # 321UPS
COMPARE
8 7/8 8 5 7/8
₹4,300.00INR / Each
ThermoSafe 324UPS Medium-Capacity Insulated Shipper, 8 x 5.8 x 11.5 in; 1/Pk 0372631 Mfr # 324UPS
COMPARE
11 1/2 8 5 7/8
₹5,400.00INR / Each
ThermoSafe 439 Medium-Capacity Insulated Shipper, 6.25 x 5.75x 7.75 in; 1/Pk 0372653 Mfr # 439
COMPARE
7 7/8 6 1/4 5 3/4
₹4,700.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat