ThermoSafe 324UPS Medium-Capacity Insulated Shipper, 8 x 5.8 x 11.5 in; 1/Pk

Representative image only.

ThermoSafe 324UPS Medium-Capacity Insulated Shipper, 8 x 5.8 x 11.5 in; 1/Pk

ThermoSafe | Mfr: 324UPS| Item: 0372631

Ship nonhazardous materials

₹5,400.00INR / Each

₹5,400.00INR / Each
Product Description

Key Features

  • Add as many ice packs as you need — choose from a variety of sizes to meet your refrigeration needs.

More About this Item

Medium-capacity models have a rectangular interior and include cartons. Mailer 03726-53 has thick walls — save money on refrigerant.
Note: For shipping nonhazardous materials only.

Specifications
  • Wall Thickness (cm)3.8
  • Wall Thickness (in)1 1/2
  • Inside Height (in)11 1/2
  • Inside Length (in)8
  • Inside Width (in)5 7/8
  • Outside Width (in)9
  • Outside Length (in)11
  • Outside Height (in)14 1/2
  • Inside Height (cm)29.2
  • Inside Length (cm)20.3
  • Inside Width (cm)14.9
  • Outside Width (cm)22.9
  • Outside Length (cm)27.9
  • Outside Height (cm)36.8
  • Shipping Weight (oz)27.2
  • Shipping Weight (g)771
  • DescriptionMedium-Capacity Insulated Shipper, 8 x 5.8 x 11.5 in; 1/Pk
Reviews
Q&A