ThermoSafe 318 Medium-Capacity Insulated Shipper, 8 x 6 x 4.25 in; 1/Pk

Representative image only.

ThermoSafe 318 Medium-Capacity Insulated Shipper, 8 x 6 x 4.25 in; 1/Pk

ThermoSafe | Mfr: 318| Item: 0372611

Ship nonhazardous materials

₹4,000.00INR / Each

₹4,000.00INR / Each
Product Description

Key Features

  • Add as many ice packs as you need — choose from a variety of sizes to meet your refrigeration needs.

More About this Item

Medium-capacity models have a rectangular interior and include cartons. Mailer 03726-53 has thick walls — save money on refrigerant.
Note: For shipping nonhazardous materials only.

Specifications
  • Wall Thickness (cm)3.8
  • Wall Thickness (in)1 1/2
  • Inside Height (in)4 1/4
  • Inside Length (in)8
  • Inside Width (in)6
  • Outside Width (in)9
  • Outside Length (in)11
  • Outside Height (in)7 1/4
  • Inside Height (cm)10.8
  • Inside Length (cm)20.3
  • Inside Width (cm)15.2
  • Outside Width (cm)22.9
  • Outside Length (cm)27.9
  • Outside Height (cm)18.4
  • Shipping Weight (oz)20.8
  • Shipping Weight (g)590
  • DescriptionMedium-Capacity Insulated Shipper, 8 x 6 x 4.25 in; 1/Pk
Reviews
Q&A