ThermoSafe 439 Medium-Capacity Insulated Shipper, 6.25 x 5.75x 7.75 in; 1/Pk

Representative image only.

ThermoSafe 439 Medium-Capacity Insulated Shipper, 6.25 x 5.75x 7.75 in; 1/Pk

ThermoSafe | Mfr: 439| Item: 0372653

Ship nonhazardous materials

₹4,700.00INR / Each

₹4,700.00INR / Each
Product Description

Key Features

  • Add as many ice packs as you need — choose from a variety of sizes to meet your refrigeration needs.

More About this Item

Medium-capacity models have a rectangular interior and include cartons. Mailer 03726-53 has thick walls — save money on refrigerant.
Note: For shipping nonhazardous materials only.

Specifications
  • Wall Thickness (cm)5.7
  • Wall Thickness (in)2 1/4
  • Inside Height (in)7 7/8
  • Inside Length (in)6 1/4
  • Inside Width (in)5 3/4
  • Outside Width (in)10 1/2
  • Outside Length (in)11 1/4
  • Outside Height (in)12 5/8
  • Inside Height (cm)20
  • Inside Length (cm)15.9
  • Inside Width (cm)14.6
  • Outside Width (cm)26.7
  • Outside Length (cm)28.6
  • Outside Height (cm)32.1
  • Shipping Weight (oz)24
  • Shipping Weight (g)680
  • DescriptionMedium-Capacity Insulated Shipper, 6.25 x 5.75x 7.75 in; 1/Pk
Reviews
Q&A