ThermoSafe Brands Large-Capacity Shippers

Ship nonhazardous materials without worry
Large-capacity models have a rectangular interior and include cartons.
Note: For shipping nonhazardous materials only.
5,800.00 - ₹17,300.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Inside Height (in) Inside Length (in) Inside Width (in)
Availability Pricing
ThermoSafe 325UP5 Large-Capacity Shipper, 11.6 x 9.8 x 7 in; 1/Pk 0373601 Mfr # 325UP5
COMPARE
7 11 5/8 9 7/8
₹5,800.00INR / Each
ThermoSafe 326 Large-Capacity Shipper, 11.6 x 9.8 x 9 in; 1/Pk 0373801 Mfr # 326
COMPARE
9 11 5/8 9 7/8
₹6,300.00INR / Each
ThermoSafe 398 Large-Capacity Shipper, 19.4 x 12.4 x 12.25 in; 1/Pk 0373900 Mfr # 398
COMPARE
12 1/4 19 3/8 12 3/8
₹17,300.00INR / Each
ThermoSafe 314 Lab-Grade Insulated Container, 10.5 x 7.25 x 10 in; 1/Pk 0374000 Mfr # 314
COMPARE
10 10 1/2 7 1/4
₹7,900.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat