ThermoSafe 398 Large-Capacity Shipper, 19.4 x 12.4 x 12.25 in; 1/Pk

Representative image only.

ThermoSafe 398 Large-Capacity Shipper, 19.4 x 12.4 x 12.25 in; 1/Pk

ThermoSafe | Mfr: 398| Item: 0373900

Ship nonhazardous materials without worry

₹17,300.00INR / Each

₹17,300.00INR / Each
Product Description

More About this Item

Large-capacity models have a rectangular interior and include cartons.
Note: For shipping nonhazardous materials only.

Specifications
  • Wall Thickness (cm)3.8
  • Wall Thickness (in)1 1/2
  • Inside Height (in)12 1/4
  • Inside Length (in)19 3/8
  • Inside Width (in)12 3/8
  • Outside Width (in)15 3/8
  • Outside Length (in)22 3/8
  • Outside Height (in)15 3/8
  • Inside Height (cm)31.1
  • Inside Length (cm)49.2
  • Inside Width (cm)31.4
  • Outside Width (cm)39.1
  • Outside Length (cm)56.8
  • Outside Height (cm)39.1
  • Shipping Weight (oz)65.6
  • Shipping Weight (g)1860
  • DescriptionLarge-Capacity Shipper, 19.4 x 12.4 x 12.25 in; 1/Pk
Reviews
Q&A