ThermoSafe 326 Large-Capacity Shipper, 11.6 x 9.8 x 9 in; 1/Pk

Representative image only.

ThermoSafe 326 Large-Capacity Shipper, 11.6 x 9.8 x 9 in; 1/Pk

ThermoSafe | Mfr: 326| Item: 0373801

Ship nonhazardous materials without worry

₹6,300.00INR / Each

₹6,300.00INR / Each
Product Description

More About this Item

Large-capacity models have a rectangular interior and include cartons.
Note: For shipping nonhazardous materials only.

Specifications
  • Wall Thickness (cm)3.8
  • Wall Thickness (in)1 1/2
  • Inside Height (in)9
  • Inside Length (in)11 5/8
  • Inside Width (in)9 7/8
  • Outside Width (in)13 1/4
  • Outside Length (in)15
  • Outside Height (in)12
  • Inside Height (cm)22.9
  • Inside Length (cm)29.5
  • Inside Width (cm)25.1
  • Outside Width (cm)33.7
  • Outside Length (cm)38.1
  • Outside Height (cm)30.5
  • Shipping Weight (oz)33.6
  • Shipping Weight (g)952
  • DescriptionLarge-Capacity Shipper, 11.6 x 9.8 x 9 in; 1/Pk
Reviews
Q&A