ThermoSafe 314 Lab-Grade Insulated Container, 10.5 x 7.25 x 10 in; 1/Pk

Representative image only.

ThermoSafe 314 Lab-Grade Insulated Container, 10.5 x 7.25 x 10 in; 1/Pk

ThermoSafe | Mfr: 314| Item: 0374000

Ship nonhazardous materials without worry

₹7,900.00INR / Each

₹7,900.00INR / Each
Product Description

More About this Item

Large-capacity models have a rectangular interior and include cartons.
Note: For shipping nonhazardous materials only.

Specifications
  • Wall Thickness (cm)3.8
  • Wall Thickness (in)1 1/2
  • Inside Height (in)10
  • Inside Length (in)10 1/2
  • Inside Width (in)7 1/4
  • Outside Width (in)10 1/2
  • Outside Length (in)13 3/4
  • Outside Height (in)13 1/2
  • Inside Height (cm)25.4
  • Inside Length (cm)26.7
  • Inside Width (cm)18.4
  • Outside Width (cm)26.7
  • Outside Length (cm)34.9
  • Outside Height (cm)34.3
  • Shipping Weight (oz)96
  • Shipping Weight (g)2721
  • DescriptionLab-Grade Insulated Container, 10.5 x 7.25 x 10 in; 1/Pk
Reviews
Q&A