Thermo Scientific Stirring Bars
  • Super-strong samarium-cobalt (SmCo) magnets provide superior mixing action at high speeds
  • Chemically inert, low-friction, leakproof PTFE coating provides smooth spinning and maximum energy transfer
  • Steam steralize to 121° C
  • Choose stir bars with glide-ring for reduced noise and higher wear resistance
16,700.00 - ₹64,500.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in)
Availability Pricing
Thermo Scientific 50087902 Komet Stir Bar 90 (Length: 3 1/2" (90mm)) WW-04651-21 Mfr # 50087902
COMPARE
1 3 1/2
₹64,500.00INR / Each
Thermo Scientific 50087909 Komet 50 Stirring Bar WW-04651-22 Mfr # 50087909
COMPARE
0.8 2
₹16,700.00INR / Each
Thermo Scientific 50087930 Komet 30 Stir Bars 5/pk WW-04651-23 Mfr # 50087930
COMPARE
0.5 1
₹42,000.00INR / Pkg of 5
Thermo Scientific 50087924 Komet 15 Stir Bars 5/pk WW-04651-24 Mfr # 50087924
COMPARE
0.3 19/32
₹29,300.00INR / Pkg of 5